Hach Gelex Secondary Turbidity Standard Kit, 0 to 4000 NTU
Representative image only.
Hach Gelex Secondary Turbidity Standard Kit, 0 to 4000 NTU
Hach | Mfr: 2589000| Item: 9951222
Optimize water analysis
$664.50CAD / Each
$664.50CAD / Each
Buy It Together
Current Item
Hach Gelex  Secondary Turbidity Standard Kit, 0 to 4000 NTU
Hach Gelex Secondary Turbidity Standard Kit, 0 to 4000 NTU
9951222
$664.50CAD / Each
+
Hach 2635300 Speccheck Standards, DPD for Chlorine
9957496
$444.96CAD / Each
+ +
Hach TL2310 LED Turbidimeter, ISO, 0 to 1000 NTU
9951220
$6,375.36CAD / Each
+
Hach 2100Q  Handheld Turbidity Meter, EPA Compliant
9951110
$3,166.32CAD / Each
+ =
Total Price 11181.86 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Gelex Secondary Standards Kit includes 1 sealed vial each, Stray Light Standard, 0-2 NTU, 0-20 NTU, 0-200 NTU, 200-4000 NTU.
Key Features
  • Choose from a variety of accessories
  • Secondary Turbidity Standards
  • 1 year warranty
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
1 Year Warranty
Specifications
  • Compatible WithHach Turbidity meters
  • DescriptionGelex secondary turbidity standard kit for TL23 series, 0 to 4000 NTU