Hach TL23 Benchtop Turbidimeters
Simplify testing in demanding industrial and wastewater applications
  • Identifies stable readings to ensure accurate analysis
  • Displays measurements more clearly on large, color touch screen
  • Locks in measurements and logs up to 2000 data points
  • Easily exports data via a USB port
The TL23 series benchtop turbidimeters simplify the measurement of high range turbidity applications with an easy-to-use color touch screen and intuitive user interface. Features include self-diagnostics for quality assurance, single-point calibration for measurements under 40 NTU/FNU, and sample identification for traceability.
Stable readings and accurate analysis are ensured by capturing turbidity reading once the turbidimeter detects sample stability. No need for repeat measurements.
$5,715.28 - $9,114.39CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Turbidity Turbidity Resolution Description
Availability Pricing
Hach TL2310 LED Turbidimeter, ISO, 0 to 1000 NTU RK-99512-20 Mfr # LPV444.99.00120
COMPARE
1000 0.001 NTU/EBC LED Turbidimeter, ISO, 0 to 1000 NTU
$5,787.48CAD / Each
Hach TL2300 Tungesten Lamp Turbidimeter, EPA, 0 to 4000 NTU RK-99512-25 Mfr # LPV444.99.00210
COMPARE
4000 0.001 NTU/EBC Tungesten Lamp Turbidimeter, EPA, 0 to 4000 NTU
$5,715.28CAD / Each
Hach TL2350 Tungesten Lamp Turbidimeter, EPA, 0 to 10,000 NTU RK-99512-30 Mfr # LPV444.99.00310
COMPARE
10000 0.001 NTU/EBC Tungesten Lamp Turbidimeter, EPA, 0 to 10,000 NTU
$9,114.39CAD / Each
Hach TL2360 LED Turbidimeter, ISO, 0 to 10,000 NTU RK-99512-35 Mfr # LPV444.99.00320
COMPARE
10000 0.001 NTU/EBC LED Turbidimeter, ISO, 0 to 10,000 NTU
$8,996.52CAD / Each