Hach 2635300 Speccheck Standards, DPD for Chlorine
Representative image only.
Hach 2635300 Speccheck Standards, DPD for Chlorine
Hach | Mfr: 2635300| Item: UZ-99574-96
Ideal for Pocket Colorimeter II calibration
$444.96CAD / Each
$444.96CAD / Each
Buy It Together
Current Item
Hach 2635300 Speccheck Standards, DPD for Chlorine
Hach 2635300 Speccheck Standards, DPD for Chlorine
9957496
$444.96CAD / Each
+
Hach 20849-00 Lab Turbidimeter Sample Cells; 6/Pk
9951270
$156.18CAD / Each
+
Hach DR 900 Multiparameter Colorimeter, Handheld
9957310
$3,630.43CAD / Each
=
Total Price 4231.57 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Four vials of known concentration standards
  • 1 year warranty
More About this Item
Spec Gel Secondary Standard kit--four sealed vials of known concentration from 0 to 2.0 mg/l for Chlorine.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • Test ParametersChloride
  • DescriptionSpeccheck Standards, DPD for Chlorine