Hach 47076-00 Lab Turbidimeter Sample Cell Oiling Cloth
Representative image only.
Hach 47076-00 Lab Turbidimeter Sample Cell Oiling Cloth
Hach | Mfr: 4707600| Item: 9951272
Optimize water analysis
$44.05CAD / Each
$44.05CAD / Each
Buy It Together
Current Item
Hach 47076-00 Lab Turbidimeter Sample Cell Oiling Cloth
Hach 47076-00 Lab Turbidimeter Sample Cell Oiling Cloth
9951272
$44.05CAD / Each
+
Hach 2635300 Speccheck Standards, DPD for Chlorine
9957496
$444.96CAD / Each
+ +
Hach TL2310 LED Turbidimeter, ISO, 0 to 1000 NTU
9951220
$6,375.36CAD / Each
+
Hach 2100Q  Handheld Turbidity Meter, EPA Compliant
9951110
$3,166.32CAD / Each
+ =
Total Price 10561.41 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
Use oiled cloth to wipe down sample cells. Oil fills in small scratches on the sample cell for more accurate readings.
Specifications
  • Compatible WithHach Meter
  • DescriptionLab Turbidimeter Sample Cell Oiling Cloth