Hach 43975-00 Turbidity Sample Degassing Kit
Representative image only.
Hach 43975-00 Turbidity Sample Degassing Kit
Hach | Mfr: 4397500| Item: 9951268
Optimize water analysis
$95.55CAD / Each
$95.55CAD / Each
Buy It Together
Current Item
Hach 43975-00 Turbidity Sample Degassing Kit
Hach 43975-00 Turbidity Sample Degassing Kit
9951268
$95.55CAD / Each
+ +
Hach 2100Q  Handheld Turbidity Meter, EPA Compliant
9951110
$3,166.32CAD / Each
+
Hach TL2310 LED Turbidimeter, ISO, 0 to 1000 NTU
9951220
$6,375.36CAD / Each
+ +
Hach 2635300 Speccheck Standards, DPD for Chlorine
9957496
$444.96CAD / Each
=
Total Price 10612.91 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
Kit includes plastic syringe with male slip tip luer and one-holed rubber stoppers to fit 2100 series turbidity meter vials.
Use is simple. Place sample into vial, put supplied rubber stopper on to the vial, insert syringe (with plunger compressed) into hole of stopper, pull up syringe plunger in order to pull a vacuum on the contents of the vial, hold for 10 seconds, remove syringe, and test sample.
Specifications
  • Compatible WithHach Meter
  • DescriptionTurbidity Sample Degassing Kit