Hach 26621-05 Turbidity Standards Calibration Kit, 0-4000 NTU; Set of 5 Standards
Representative image only.
Hach 26621-05 Turbidity Standards Calibration Kit, 0-4000 NTU; Set of 5 Standards
Hach | Mfr: 2662105| Item: 9951250
Optimize water analysis
$574.62CAD / Each
$574.62CAD / Each
Buy It Together
Current Item
Hach 26621-05 Turbidity Standards Calibration Kit, 0-4000 NTU; Set of 5 Standards
Hach 26621-05 Turbidity Standards Calibration Kit, 0-4000 NTU; Set of 5 Standards
9951250
$574.62CAD / Each
+ +
Hach 2100Q  Handheld Turbidity Meter, EPA Compliant
9951110
$3,166.32CAD / Each
+
Hach TL2310 LED Turbidimeter, ISO, 0 to 1000 NTU
9951220
$6,375.36CAD / Each
+ +
Hach 2635300 Speccheck Standards, DPD for Chlorine
9957496
$444.96CAD / Each
=
Total Price 11091.98 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Includes
sealed vials of <0.1, 20, 200, 1000 and 4000 NTU standards.
Key Features
  • Choose from a variety of accessories
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
Specifications
  • Compatible WithHACH 2100AN turbidimeter, 99512-00 & 99512-07
  • DescriptionCalibration sets for Laboratory Turbidimeter, item numbers 99512-00, -05