Hach 21258-25 COD Test Kit, Low Range, 0 to 150 ppm
Representative image only.
Hach 21258-25 COD Test Kit, Low Range, 0 to 150 ppm
Hach | Mfr: 2125825| Item: 9957425
Optimize water analysis
$99.45CAD / Each
$99.45CAD / Each
Buy It Together
Current Item
Hach 21258-25 COD Test Kit, Low Range, 0 to 150 ppm
Hach 21258-25 COD Test Kit, Low Range, 0 to 150 ppm
9957425
$99.45CAD / Each
+ +
Hach 2100Q  Handheld Turbidity Meter, EPA Compliant
9951110
$3,166.32CAD / Each
+
Hach TL2310 LED Turbidimeter, ISO, 0 to 1000 NTU
9951220
$6,375.36CAD / Each
+ +
Hach 2635300 Speccheck Standards, DPD for Chlorine
9957496
$444.96CAD / Each
=
Total Price 10616.81 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
  • Use with Hach colorimeter and COD reactor (below)
  • 1 year warranty
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
Kits contain leak-proof vials. Caps provide a positive seal that traps volatile material and ensures complete digestion. Reagents are premixed, minimizing dangerous spills and handling of corrosive and toxic chemicals. Small reagent volumes mean less silver and mercury waste. Pack of 25 reagent vials.
Canadian Standards Association1 Year WarrantyEPA United States Environmental protection agencyUL Listed.
Specifications
  • Compatible WithHACH DR/870
  • DescriptionCOD Test Kit, Low Range, 0 to 150 ppm