Environmental Express 1500 pH Benchtop Meter Kit

Representative image only.

Environmental Express 1500 pH Benchtop Meter Kit

Environmental Express | Item: 3541940

Optimized for laboratory measurements

$1,060.84CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,060.84CAD / Each

Buy It Together

Current Item

Product Description

Key Features

  • Large monochrome LCD
  • Integrated electrode holder (up to 2 electrodes) that can be attached to either side of the meter
  • Simple user interface and single parameter display
  • 1000 Memory data points
  • Automatic Temperature Compensation (ATC) with temperature calibration
  • Adjustable auto shut-off time (1 to 30 minutes)
  • Auto stable, auto hold, and real time measurement modes with reading stability indicators
  • PC (standard USB) / printer (25 pin serial) connection via 2.5 mm diameter phono jack
  • 3 year warranty

More About this Item

pH models measure pH and ORP/mV readings. In addition to measurement parameters, each model can be bought as a kit or a meter only (meter only versions include the integrated electrode stand).

3 Year WarrantyConformite Europeenne
Specifications
  • Meter TypeBenchtop
  • Display typeStatic segment LCD with 160 segments
  • ConnectionsBNC, phono (ATC), DC sockets
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01 pH
  • pH Accuracy± 0.01 pH
  • Buffer RecognitionYes
  • CalibrationYes
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 mV (< ±1000 mV), 1 mV (≥ ±1000 mV)
  • mV Accuracy±0.3 mV (< ±1000 mV), 0.3% of reading (≥ ±1000 mV)
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1 °C / °F
  • Temperature Accuracy± 0.5 °C / ± 0.9 °F
  • Temperature compensationYes
  • OutputPhono jack (to USB / RS-232)
  • Data Logging (points)1000 data points
  • Memory Size1000 data points
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Number of Positions2
  • Probe IncludedYes
  • DescriptionpH Benchtop Meter Kit
  • Warranty3 year
Reviews
Q&A