Environmental Express Cole Parmer P200-02 pH Meter Kit; 3-in-1 pH Electrode, Electrode Holder

Representative image only.

Environmental Express Cole Parmer P200-02 pH Meter Kit; 3-in-1 pH Electrode, Electrode Holder

Environmental Express | Item: 5925005

Options for every application

$1,018.11CAD / Each

$1,018.11CAD / Each

Buy It Together

Current Item

Environmental Express Cole Parmer P200-02 pH Meter Kit; 3-in-1 pH Electrode, Electrode Holder
Environmental Express Cole Parmer P200-02 pH Meter Kit; 3-in-1 pH Electrode, Electrode Holder
5925005

$1,018.11CAD / Each
+ + + + +
Environmental Express 1500 pH Benchtop Meter Kit
3541940

$1,060.84CAD / Each
+ + + =
Total Price 6127.67 ADD 9 ITEMS TO CART
Product Description

Includes

flexible-arm electrode holder, pH electrode, and power cord.

Key Features

  • Choice of electrodes—combination 3-in-1 with ATC or Tris-compatible
  • Flexible-arm electrode holder lets you perform other tasks while waiting for readings to stabilize

More About this Item

The P200 series of benchtop meters provides accurate, reliable results with the benefit of convenient accessories to make your lab run smoothly. Large color TFT display is easily visible, and features a stability icon to alert you when your readings are ready to be recorded and an electrode health icon indicating when you need to refill or replace your electrode.

3 Year Warranty
Specifications
  • Meter TypeBenchtop
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01, 0.1
  • pH Accuracy±0.01
  • Buffer RecognitionUSA, NIST, user defined
  • Calibration1 to 3 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Accuracy±0.01% full-scale
  • Max Conductivity
  • Min Temperature (° C)0
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1°F/C
  • Temperature Accuracy±0.36°F (0.2°C)
  • Temperature compensationYes, 32 to 212°F (0 to 100°C), automatic or manual
  • Power (VAC)110 - 120
  • Power (Hz)60
  • DescriptionP200-02 pH Meter Kit; 3-in-1 pH Electrode, Electrode Holder
Reviews
Q&A