Environmental Express 1500 PC Benchtop Meter with Electrode Stand

Representative image only.

Environmental Express 1500 PC Benchtop Meter with Electrode Stand

Environmental Express | Item: 3541951

Optimized for laboratory measurements such as pH, ORP/mV, conductivity, resistivity, TDS, salinity, and temperature

$1,502.86CAD / Each

$1,502.86CAD / Each

Buy It Together

Current Item

Environmental Express 1500 PC Benchtop Meter with Electrode Stand
Environmental Express 1500 PC Benchtop Meter with Electrode Stand
3541951

$1,502.86CAD / Each
+ +
Environmental Express 1500 pH Benchtop Meter Kit
3541940

$1,060.84CAD / Each
+
Environmental Express 1500 All-in-One pH Electrode
3541942

$278.47CAD / Each
=
Total Price 3857.34 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Meter, electrode stand, and power supply. Electrodes or calibration solutions sold separately.

Key Features

  • Large monochrome LCD
  • Integrated electrode holder (up to 2 electrodes) that can be attached to either side of the meter
  • Simple user interface and single parameter display
  • 1000 Memory data points
  • Automatic Temperature Compensation (ATC) with temperature calibration
  • Adjustable auto shut-off time (1 to 30 minutes)
  • Auto stable, auto hold, and real time measurement modes with reading stability indicators
  • PC (standard USB) / printer (25 pin serial) connection via 2.5 mm diameter phono jack
  • Probes not included must be purchased seperately 35419-42 pH & 35419-47 conductivity cell
  • 3 year warranty

More About this Item

Measurement parameters available vary by model. EC models measure conductivity, TDS, salinity, and resistivity. PC models measure pH, ORP/mV, conductivity, TDS, salinity, and resistivity. Users can choose the part number that best fits their needs. In addition to measurement parameters, each model can be bought as a kit or a meter only (meter only versions include the integrated electrode stand).

3 Year WarrantyConformite Europeenne
Specifications
  • Meter TypeBenchtop
  • Display typeStatic segment LCD with 160 segments
  • ConnectionsBNC, phono (ATC), DC sockets
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01 pH
  • pH Accuracy± 0.01 pH
  • Buffer RecognitionYes
  • CalibrationYes
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 mV (< ±1000 mV), 1 mV (≥ ±1000 mV)
  • mV Accuracy±0.3 mV (< ±1000 mV), 0.3% of reading (≥ ±1000 mV)
  • Max Conductivity0.00 – 20.00 μS, 20.1 – 200.0 μS, 201– 2000 μS, 2.01 – 20.00 mS, 20.1 – 200.0 mS
  • Conductivity Resolution0.01 / 0.1
  • Conductivity Accuracy±0.6% full scale; ±1.5% full scale > 18.0 mS/cm
  • Max TDS0 – 10.00 ppm, 10.1 – 100.0 ppm, 101 – 1000 ppm, 1.01 – 10.00 ppt, 10.1 – 100 ppt
  • TDS Resolution0.01 / 0.1 / 1 ppm ; 0.01 / 0.1 ppt
  • TDS Accuracy±0.1% full scale
  • Min Salinity0.0 ppt / 0.00%
  • Max Salinity100.0 ppt / 10.00 %
  • Salinity Resolution0.1 ppt / 0.01%
  • Salinity Accuracy±0.2% full scale
  • Min Resistivity0.000 Ω•cm
  • Max Resistivity 20.0 MΩ•cm
  • Resistivity Resolution0.5% full scale
  • Resistivity Accuracy±0.6% full scale, ±1.5% full scale > 1.80 MΩ•cm
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1 °C / °F
  • Temperature Accuracy± 0.5 °C / ± 0.9 °F
  • Temperature compensationYes
  • OutputPhono jack (to USB / RS-232)
  • Data Logging (points)1000 data points
  • Memory Size1000 data points
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Number of Positions2
  • Accessories IncludedElectrode stand
  • DescriptionPC Benchtop Meter with Electrode Stand
  • Warranty3 year
Reviews
Q&A