This item is now discontinued. Please browse similar items
Davis Instruments 6153 Wireless Weather Station with 24-Hour Fan Aspirated Radiation Shield
Representative image only.
Davis Instruments Mfr # 6153 – Item # RK-34542-65
Versatile wireless weather station complete with rain collector, temperature/humidity sensors and anemometer
  • Wireless integrated sensor suite runs on solar power; lithium battery provides backup at night and during cloudy, wintry days
  • Rain bucket is self-emptying for exceptionally accurate readings
  • Temperature and humidity sensors are located inside the radiation protective shield
  • Sophisticated forecasting algorithm takes wind and temperature into account - not just barometric pressure
  • Over 80 graphs and over 70 alarms available on the console
  • Anemometer is detachable for ultimate flexibility - mount with integrated sensor suite or mount separately using the included 40 ft cable
  • Optional tripod makes installation easy - tilts to mount on roof or uneven terrain
  • Includes console, integrated sensor suite, radiation fan/shield, and mounting hardware
SPECS & DESCRIPTION
Specifications & Description
  • Max Wind Speed (mph)200
  • Max Temperature (Outside) (° F)150
  • Max Temperature (Outside) (° C)65
  • Max Temperature (Inside) (° F)140
  • Max Temperature (Inside) (° C)60
  • Dew Point Accuracy±3°F (1.5°C)
  • Wind Speed Accuracy±5%
  • Max Wind Chill (° F)135
  • Max Wind Chill (° C)57
  • Wind Chill Accuracy±2°F (1°C)
  • Display TypeLCD
  • DescriptionWireless Weather Station with 24-Hour Fan Aspirated Radiation Shield
Key Features
  • Wireless integrated sensor suite runs on solar power; lithium battery provides backup at night and during cloudy, wintry days
  • Rain bucket is self-emptying for exceptionally accurate readings
  • Temperature and humidity sensors are located inside the radiation protective shield
  • Sophisticated forecasting algorithm takes wind and temperature into account - not just barometric pressure
  • Over 80 graphs and over 70 alarms available on the console
  • Anemometer is detachable for ultimate flexibility - mount with integrated sensor suite or mount separately using the included 40 ft cable
  • Optional tripod makes installation easy - tilts to mount on roof or uneven terrain
  • Includes console, integrated sensor suite, radiation fan/shield, and mounting hardware
More About this Item
This powerful station transmits data up to 1000 ft or even further with an optional wireless repeater. Measured parameters update every 2.5 seconds as compared to 3 to 15 minutes found on other stations.
Available for sale in the United States and Canada only.