Davis Instruments 7716 Mounting Tripod for Weather Stations, 5.8 ft tall
Representative image only.
Davis Instruments 7716 Mounting Tripod for Weather Stations, 5.8 ft tall
Davis Instruments | Mfr: 7716| Item: 8640370
Brackets at the base of each leg tilt to mount on your roof or uneven terrain
$358.92CAD / Each
$358.92CAD / Each
Product Description
Includes
two 3-foot (0.92-meter) poles and mounting hardward
Key Features
  • Galvanized steel construction
  • 1 year warranty
More About this Item
Use a mounting tripod for your Davis Vantage Pro2 or Vantage Vue to make installation even easier. Includes two 3-foot (0.92-meter) poles, which may be used separately or together to make a single 5.8-foot (1.77-meter) pole.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • DescriptionMounting Tripod for Weather Stations, 5.8 ft tall