Davis Instruments Anemometer/Sensor Transmitter Kit
Representative image only.
Davis Instruments Anemometer/Sensor Transmitter Kit
Davis Instruments | Mfr: 6332| Item: 3000362
Take air measurement readings from a more favorable location without moving the main integrated sensor suite.
$940.61CAD / Each
for an additional
$604.09CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$940.61CAD / Each
Buy It Together
Current Item
Davis Instruments Anemometer/sensor transmitter kit
Davis Instruments Anemometer/sensor transmitter kit
3000362
$940.61CAD / Each
+ + + =
Total Price 2085.29 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
transmitter inside a weather-resistant shelter, solar panel, battery and mounting hardware.
Key Features
  • Install additional sensors in different locations
  • 1 year warranty
More About this Item
Use this anemometer/sensor transmitter kit only with Vantage Pro2 or Vantage Vue weather station console/receivers. The unit is solar-powered, but has secondary, backup power coming from a 3 V lithium battery or optional Vantage Pro2 AC power adapter.
The kit can transmit up to 1,000 feet (line of sight) or 200 to 400 feet through walls and other obstacles to your wireless console/receiver. The transmitter can also accept signal from solar radiation, UV, temperature, and rain sensors.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • DescriptionAnemometer/sensor transmitter kit