Atago 2612 Refractive Index Refractometer, 1.435 to 1.520 Refractive Index
Representative image only.
Atago 2612 Refractive Index Refractometer, 1.435 to 1.520 Refractive Index
Atago | Mfr: 2612| Item: 0294009
Units provide high precision, accurate results
$691.98CAD / Each
for an additional
$200.01CAD / Each
for an additional
$321.94CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$691.98CAD / Each
Buy It Together
Current Item
Atago 2612 Refractive Index Refractometer, 1.435 to 1.520 Refractive Index
Atago 2612 Refractive Index Refractometer, 1.435 to 1.520 Refractive Index
0294009
$691.98CAD / Each
+ +
InnoCal NIST-Traceable Calibration; Refractometer
1710500
$200.01CAD / Each
+ =
Total Price 2559.14 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measure RI of stock solutions, dilutions, oils, and petrochemical products
  • Survey oil slicks and spills
  • 1 year warranty
More About this Item
Model 02940-09 features a rubber-insulated grip. Model 02940-12 has three switch-selectable ranges for high readability over entire range.
Model 02940-09 features a rubber-insulated grip.
1 Year Warranty
Specifications
  • Min Refractive Index1.43
  • Max Refractive Index1.52
  • Resolution0.001 (RI)
  • Accuracy±0.001 (RI)
  • DescriptionRefractive Index Refractometer, 1.435 to 1.520 Refractive Index