ATAGO® Refractive Index Refractometers
Units provide high precision, accurate results
  • Measure RI of stock solutions, dilutions, oils, and petrochemical products
  • Survey oil slicks and spills
Model 02940-09 features a rubber-insulated grip. Model 02940-12 has three switch-selectable ranges for high readability over entire range.
$691.98 - $2,337.61CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Refractive Index Interval Resolution
Availability Pricing
Atago 2612 Refractive Index Refractometer, 1.435 to 1.520 Refractive Index 0294009 Mfr # 2612
COMPARE
±0.001 (RI) 0.001 (RI)
$691.98CAD / Each
Atago 2350 R5000 Refractive Index Refractometer, 1.333 to 1.520 Refractive Index 0294012 Mfr # 2350 R5000
COMPARE
±0.001 (RI) 0.001 (RI)
$2,337.61CAD / Each