AccuMac AM8010 Handheld Precision Thermometer
Representative image only.
AccuMac AM8010 Handheld Precision Thermometer
AccuMac | Mfr: AM8010| Item: 9045274
High accuracy, fast readings and great stability
$2,824.49CAD / Each
$2,824.49CAD / Each
Buy It Together
Current Item
AccuMac AM8010 Handheld Precision Thermometer
AccuMac AM8010 Handheld Precision Thermometer
9045274
$2,824.49CAD / Each
+ + + =
Total Price 13316.50 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
The AccuMac AM8010 is a handheld precision thermometer with high accuracy, fast readings and great stability. This readout is an excellent choice for field applications as well as lab measurements. Though small in size,
The AM8010 is powerful when it comes to accuracy, which is as high as 0.03 °C at 0 °C. A USB interface allows user to log and display real time data on a PC.
AM8010 allows users to choose ITS-90, IEC-751 (DIN), or Callendar-Van Dusen conversion methods to response various PRTs. All probe constants and coefficients can be keyed in through front panel.
1 Year Warranty
Specifications
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)1562
  • Min Temperature (° C)-200
  • Max Temperature (° C)850
  • DescriptionHandheld Precision Thermometer