Fluke Calibration 1524 Handheld Thermometer with Datalogger Model
Representative image only.
Fluke Calibration 1524 Handheld Thermometer with Datalogger Model
Fluke Calibration | Mfr: 1524-156| Item: 9320251
The first standards thermometers to fit into the palm of your hand.
$4,287.56CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$4,287.56CAD / Each
Buy It Together
Current Item
Fluke Calibration 1524 Handheld Thermometer with Datalogger Model
Fluke Calibration 1524 Handheld Thermometer with Datalogger Model
9320251
$4,287.56CAD / Each
+ +
AccuMac AM8010 Handheld Precision Thermometer
9045274
$2,824.49CAD / Each
=
Total Price 7215.19 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
NIST traceable certificate of calibration, CD-ROM users guide, RS-232 serial cable, software for computer interface, power supply, and 3 AA batteries.
Key Features
  • Visual trending and measurement
  • Simultaneously measures two channels
  • Reads PRTs, thermocouples, and thermistors
  • “Plug-and-play” calibration coefficients with SMART LEMO connector
  • Trends data graphically
  • Documents readings and statistics
  • 1 year warranty
More About this Item
The 1524 model is not just basic hand held thermometers, but in fact a highly precise reference thermometer. This rugged thermometer is designed for engineers and technicians that want accuracy and versatility in an easy to use package. Use with RTD's (PRTs), thermocouples, and thermistors probes to read, document, and visually trend temperature data.
The unit uses a smart LEMO connector that automatically loads calibration coefficients unique to each probe for easy and accurate plug and play operation (Only Fluke calibration probes with a "P" style Info-Con connector will fit this meter.) .
Includes a free software program that makes it easy to download logged data and program new calibration coefficients.
Available for sale in the United States and Canada only.
Model 1524
Dual channels that reads RTD's (PRTs), thermocouples, and thermistors. Has all of the features of the 1523 but also logs up to 15,000 data points with a time and date stamp.
Hart Scientific is now Fluke Calibration.
NIST Traceable Calibration1 Year Warranty
Specifications
  • Meter Measurement TypeMulti-Parameter
  • Number Of Probe Channels2
  • Types Of Probes AcceptedThermocouple, Thermistor, RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)3182
  • Min Temperature (° C)-200
  • Max Temperature (° C)1750
  • Accuracy0.004% + 0.002 ohm percent of reading + floor
  • Resolution0.0001°
  • Data LoggingYes
  • Memory Capacity2000 data points
  • Battery Life20 hours
  • Battery3 AA (included)
  • DescriptionHandheld Thermometer with Datalogger Model