Cole-Parmer® VapLock Solvent Waste Collection Systems and Kits
TWO EASY OPTIONS FOR ORDERING A CLOSED SOLVENT SYSTEM
Cole-Parmer VapLock Complete Solvent Waste Collection System
Or
Cole-Parmer VapLock Solvent Waste Collection Kit
Solvent Collection Systems— choose container style and material
All systems come with container, manifold, 5-feet of tubing with union connections, tubing adapters, plugs for any unused ports, and an exhaust filter with breakthrough color indicator.
Shop by manifold and container specs you need.  Compare Full Specs >
Solvent Waste Kits— use with your existing container
Waste Accessory Kits include a solvent collection manifold (Choose the kit appropriate for your container), an organic solvent exhaust filter with breakthrough indicator, and adapters, plugs, and tubing connections required for installation on typical HPLC instruments. Container not included.