Justrite Quick-Disconnect Disposal Safety Cans
Remove can and empty waste without disconnecting inlet tubing
  • Automatically vents at 3-5 psig
  • Accepts 3/8" ID tubing
  • FM approved
  • OSHA and NFPA compliant
  • EPA complaint
Polyethylene disposal safety cans provide a safe, hands-free way to drain waste liquid from your HPLC applications. Protect the liquid and vapors inside from external heat sources. Quick-disconnect are valved open when connected and valved closed when disconnected for spill free changeout of safety cans without disconnecting the tubing.
Coalescing carbon filter attaches to one of the fittings to filter VOCs. Durable polyethylene is translucent for quick visual inspection of liquid level. Pouring spout includes stainless steel flame arrester to prevent flashback ignition. Dual fittings (intake, vent) eliminate back pressure and provide controlled handling of vapors.
$729.70 - $1,166.19CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Capacity (Liters) Material
Availability Pricing
Justrite PP12752 Quick-Disconnect Disposal Safety Can, Polyethylene, 2 Gal 8639500 Mfr # PP12752
COMPARE
2 7.57 Polyethylene(PE)
$729.70CAD / Each
Justrite PP12755 Quick-Disconnect Disposal Safety Can, Polyethylene, 5 Gal 8639510 Mfr # PP12755
COMPARE
5 18.93 Polyethylene(PE)
$1,166.19CAD / Each