This product is now discontinued, View Related Products
Cole-Parmer Waste Safety Trays
Enhance the safety of your HPLC solvent waste system
  • Constructed of tough and durable HDPE
  • Designed with the capacity to hold 115% of carboy's content
Contain leakage and spills from solvent waste carboys with this secondary container. Available in single or multi-pack, and come in different capacities to accommodate a range of carboys. Red color for high visibility.
$34.99 - $409.60CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Material Description
Availability Pricing
COMPARE
14 HDPE Safety Tray for 1L to 10L Carboys; 1 Ea
$34.99CAD / Each
COMPARE
34 HDPE Safety Tray for 10L to 20L Carboys; 1 Ea
$58.94CAD / Each
COMPARE
73.4 HDPE Safety Tray for 20L to 60L Carboys; 1 Ea
$115.22CAD / Each
COMPARE
103.7 HDPE Safety Tray for 60L to 90L Carboys; 1 Ea
$138.50CAD / Each
COMPARE
14 HDPE Safety Tray for 1L to 10L Carboys; 6/Pk
$157.65CAD / Pkg of 6
COMPARE
34 HDPE Safety Tray for 10L to 20L Carboys; 6/Pk
$296.15CAD / Pkg of 6
COMPARE
73.4 HDPE Safety Tray for 20L to 60L Carboys; 3/Pk
$231.32CAD / Pkg of 3
COMPARE
103.7 HDPE Safety Tray for 60L to 90L Carboys; 3/Pk
$409.60CAD / Pkg of 3
Showing 1- 8 of 8