TSI DP-Calc Micromanometers
Simple-to-operate, handheld digital micromanometer
  • Measure differential and static pressure from –15 to 15 in. H2O (–3735 to 3735 Pa)
The DP-CALC™ micromanometers allow you to easily make fast, accurate differential and static pressure measurements. These robust instruments can also calculate and display velocity when using an optional pitot tube.
$1,298.06 - $1,966.98CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
TSI 5815 DP-Calc Micromanometer 0594929 Mfr # 5815
COMPARE
±1.5% at 2,000 ft/min (10.16 m/s) 1 ft/min (0.1 m/s) 15500
$1,298.06CAD / Each
TSI 5825 DP-Calc Micromanometer, Model 1050877 Mfr # 5825
COMPARE
±1.5% at 2,000 ft/min (10.16 m/s) 1 ft/min (0.1 m/s) 15500
$1,966.98CAD / Each