Trajan Splitless FocusLiner

Spot the Difference in the SGE Lineup!
  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
Two focus points ensure quartz wool remains in the optimum location. Quartz wool prevents droplet formation ensuring excellent reproducibility.
$51.57 - $960.65CAD / Pkg of 5

13 variations of this product are available.

See All Product Options Below
13 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 092002 Split/Splitless Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1600922 Mfr # 092002
COMPARE
$219.54CAD / Pkg of 5
Trajan SGE 09200201 Split/Splitless Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 1/Ea 1600923 Mfr # 09200201
COMPARE
$51.57CAD / Pkg of 1
Trajan SGE 092005 Splitless Fast Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1600928 Mfr # 092005
COMPARE
$248.63CAD / Pkg of 5
Trajan SGE 092008 Split/Splitless Fast Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 25/Pk 1600932 Mfr # 092008
COMPARE
$917.92CAD / Pkg of 25
Trajan SGE 092037 Focus Liners, Compatible with Varian 1078/1079, 3.4 mm ID, 54 mm L; 5/Pk 1600967 Mfr # 092037
COMPARE
$381.24CAD / Pkg of 5
Trajan SGE 092048 Split Focus Liners, 50 mm Needle, Compatible with Thermo Trace Liner, 5 mm ID; 5/Pk 1600982 Mfr # 092048
COMPARE
$368.72CAD / Pkg of 5
Trajan SGE 092049 Splitless Focus Liners, Compatible with 50 mm Needle, Compatible with Thermo Trace Liner, 5 mm ID; 5/Pk 1600983 Mfr # 092049
COMPARE
$367.98CAD / Pkg of 5
Trajan SGE 092059 Focus Liners, Compatible with Shimadzu 2010/17A, 3.4 mm ID , 95 mm L; 5/Pk 1600988 Mfr # 092059
COMPARE
$274.79CAD / Pkg of 5
Trajan SGE 09206225 Focus Liner, Caompatible with Shimadzu 17A, 3.4 mm ID, 95 mm L; 25/Pk 1600995 Mfr # 09206225
COMPARE
$960.65CAD / Pkg of 25
Trajan SGE 092092 Split/Splitless Focus Liner, Compatible with PerkinElmer Systems, 4 mm ID, 92 mm L; 5/Pk 1601027 Mfr # 092092
COMPARE
$341.46CAD / Pkg of 5
Trajan SGE 092101 Split/Splitless Focus Liner, Compatible with PerkinElmer PSS injector, 2 mm ID; 5/Pk 1601042 Mfr # 092101
COMPARE
$387.50CAD / Pkg of 5
Trajan SGE 092219 Split/Splitless Focus Liner, Compatible with Agilent Systems, 4 mm ID, 78.5 mm L; 25/Pk 1601071 Mfr # 092219
COMPARE
$861.93CAD / Pkg of 25
Trajan SGE 092272 Split/Splitless FocusLiner, Compatible with ATAS Optic Injectors, 3 mm ID, 81 mm L; 5/Pk 1601094 Mfr # 092272
COMPARE
$229.48CAD / Pkg of 5
Showing 1- 13 of 13

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat