Trajan SGE 09206225 Focus Liner, Caompatible with Shimadzu 17A, 3.4 mm ID, 95 mm L; 25/Pk

Representative image only.

Trajan SGE 09206225 Focus Liner, Caompatible with Shimadzu 17A, 3.4 mm ID, 95 mm L; 25/Pk

Trajan | Mfr: 09206225| Item: 1600995

Spot the Difference in the SGE Lineup!

$960.65CAD / Pkg of 25

$960.65CAD / Pkg of 25
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Two focus points ensure quartz wool remains in the optimum location. Quartz wool prevents droplet formation ensuring excellent reproducibility.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Focus Liner, Caompatible with Shimadzu 17A, 3.4 mm ID, 95 mm L; 25/Pk
  • Warranty6 month warranty
Reviews
Q&A