Trajan SGE 092101 Split/Splitless Focus Liner, Compatible with PerkinElmer PSS injector, 2 mm ID; 5/Pk

Representative image only.

Trajan SGE 092101 Split/Splitless Focus Liner, Compatible with PerkinElmer PSS injector, 2 mm ID; 5/Pk

Trajan | Mfr: 092101| Item: 1601042

Spot the Difference in the SGE Lineup!

$387.50CAD / Pkg of 5

$387.50CAD / Pkg of 5
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Two focus points ensure quartz wool remains in the optimum location. Quartz wool prevents droplet formation ensuring excellent reproducibility.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE PerkinElmer Split/Splitless Focus Liner for PSS injector, 2 mm ID, 86.2 mm L; 5/Pk
  • Warranty6 month warranty
Reviews
Q&A