Trajan ConnecTite Inlet Liners

Spot the Difference in the SGE Lineup!
  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
Longer life, less maintenance downtme, and reduced maintenance costs. ConnecTite liners reduce contact with hot metal components in the inlet, inhibiting sample degredation.
$569.47 - $617.35CAD / Pkg of 5

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 092324 ConnecTite Liner, Standard, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1601100 Mfr # 092324
COMPARE
$569.47CAD / Pkg of 5
Trajan SGE 092334 ConnecTite Liner, Standard, Compatible with Thermo Scientific Systems, 105 mm L; 5/Pk 1601109 Mfr # 092334
COMPARE
$617.35CAD / Pkg of 5
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat