Trajan SGE 092324 ConnecTite Liner, Standard, Compatible with Agilent Systems, 78.5 mm L; 5/Pk

Representative image only.

Trajan SGE 092324 ConnecTite Liner, Standard, Compatible with Agilent Systems, 78.5 mm L; 5/Pk

Trajan | Mfr: 092324| Item: 1601100

Spot the Difference in the SGE Lineup!

$569.47CAD / Pkg of 5

$569.47CAD / Pkg of 5
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Longer life, less maintenance downtme, and reduced maintenance costs. ConnecTite liners reduce contact with hot metal components in the inlet, inhibiting sample degredation.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Agilent ConnecTite Liner, Standard, 78.5 mm L; 5/Pk
  • Warranty6 month warranty
Reviews
Q&A