Trajan SGE 092334 ConnecTite Liner, Standard, Compatible with Thermo Scientific Systems, 105 mm L; 5/Pk

Representative image only.

Trajan SGE 092334 ConnecTite Liner, Standard, Compatible with Thermo Scientific Systems, 105 mm L; 5/Pk

Trajan | Mfr: 092334| Item: 1601109

Spot the Difference in the SGE Lineup!

$617.35CAD / Pkg of 5

$617.35CAD / Pkg of 5
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Longer life, less maintenance downtme, and reduced maintenance costs. ConnecTite liners reduce contact with hot metal components in the inlet, inhibiting sample degredation.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Thermo Scientific ConnecTite Liner, Standard, 105 mm L; 5/Pk
  • Warranty6 month warranty
Reviews
Q&A