Thermo Scientific Nalgene Sterile Media Bottles, PETG
Inexpensive alternative to glass media bottles
  • Allows long-term media storage
  • Radiation-sterilized and nonpyrogenic
  • Tamper-resistant
Durable PETG (Polyethylene Terephthalate Glycol) bottles with HDPE closures are radiation-sterilized and nonpyrogenic—eliminates washing, depyrogenation, and autoclaving steps. Reduced permeability to CO2/O2 allows long-term media storage. Tamper-resistant band around closure and neck of each bottle ensures sterility. Bottles come in shrink-wrapped trays.
ITEM(S) ON SALE
$394.87 - $683.65CAD / Box of 24
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2019-0030 Sterile PETG Media Bottles, 30 mL; 96/box 0625400 Mfr # 2019-0030
COMPARE
30 20 mm Polyethylene Terephthalate Glycol
$683.65CAD / Box of 96
Thermo Scientific Nalgene 2019-0060 Sterile PETG Media Bottles, 60 mL; 96/box 0625410 Mfr # 2019-0060
COMPARE
60 24 mm Polyethylene Terephthalate Glycol
$498.80CAD / Box of 96
$626.19CAD / Box of 96
Thermo Scientific Nalgene 2019-0125 Sterile PETG Media Bottles, 125 mL; 48/box 0625420 Mfr # 2019-0125
COMPARE
125 38-430 Polyethylene Terephthalate Glycol
$436.12CAD / Box of 48
Thermo Scientific Nalgene 2019-0250 Sterile PETG Media Bottles, 250 mL; 48/box 0625430 Mfr # 2019-0250
COMPARE
250 38-430 Polyethylene Terephthalate Glycol
$549.57CAD / Box of 48
Thermo Scientific Nalgene 2019-0500 Sterile PETG Media Bottles, 500 mL; 24/box 0625440 Mfr # 2019-0500
COMPARE
500 38-430 Polyethylene Terephthalate Glycol
$394.87CAD / Box of 24
Thermo Scientific Nalgene 2019-1000 Sterile PETG Media Bottles, 1000 mL; 24/box 0625450 Mfr # 2019-1000
COMPARE
1000 38-430 Polyethylene Terephthalate Glycol
$518.63CAD / Box of 24
Thermo Scientific Nalgene 2019-2000 Sterile PETG Media Bottles, 2000 mL; 12/cs 0625460 Mfr # 2019-2000
COMPARE
2000 53B Polyethylene Terephthalate Glycol
$452.33CAD / Case of 12
Showing 1- 7 of 7