Thermo Scientific Heratherm General Protocol (GP) Gravity Convection Ovens
Optimally suited to everyday heating and drying applications up to 250°C
  • Small footprint saves space without compromising volume
  • Gravity convection provides gentle drying and heating with low air turbulence
  • Energy efficient — up to 20% more energy savings compared to similar units
Energy-efficient gravity convection ovens feature a gentle air circulation pattern with low turbulence to ensure accurate, even heating from 50 to 250°C, perfect for working with materials that are sensitive to airflow such as powders and foils. Large easy-to-view vacuum fluorescent display with built-in timer and touch-button controls allows quick and simple temperature setting. The exhaust vent doubles as an access port for external temperature sensors.
Smooth corrosion-resistant galvanized steel inner chamber has rounded corners for easy cleaning. The door opens 180°, providing easy access to interior for cleaning and loading of probes. An automatic overtemperature alarm ensures your samples are always protected. The RS-232 interface lets you connect to your computer for documentation of parameters.
$4,434.90 - $11,669.25CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Thermo Scientific Heratherm 51028872 GP Gravity Oven; 3.75 cu ft/120V 3880028 Mfr # 51028872
COMPARE
3.75 120
$5,127.40CAD / Each
Thermo Scientific Heratherm 51028874 GP Gravity Oven; 6.2 cu ft/120V 3880029 Mfr # 51028874
COMPARE
6.2 120
$6,158.77CAD / Each
Thermo Scientific Heratherm 51028112 GP Gravity Oven; 2.0 cu ft/120V 3880030 Mfr # 51028112
COMPARE
2.3 120
$4,434.90CAD / Each
Thermo Scientific Heratherm 51028113 GP Gravity Oven; 3.5 cu ft/208-240V 3880032 Mfr # 51028113
COMPARE
3.75 208 / 240
$5,127.40CAD / Each
Thermo Scientific Heratherm 51029327 GP Gravity Oven; 14.8 cu ft/208-240V 3880033 Mfr # 51029327
COMPARE
14.8 208 / 240
$11,669.25CAD / Each