Thermo Scientific Heratherm Advanced Protocol (AP) Mechanical Convection Ovens
Maintain even and accurate heating
  • Mechanical convection provides fast sample drying and heating with superior temperature uniformity
  • Energy efficient — up to 20% more energy savings compared to similar units
  • Small footprint saves space without compromising volume
  • Store up to 10 programs with 10 discrete steps
Advanced protocol mechanical convection ovens feature superior temperature uniformity to ensure accurate, even heating from 50 to 330°C — perfect for drying, testing, or curing materials that require the highest temperature accuracy.
Program controller with large easy-to-view display and touch-button controls. Easily set the adjustable fan speed depending on your application — highest for best temperature stability and uniformity, lowest for minimal airflow. Unique timer allows you to automatically turn the oven on or off at a preset time or operate on a recurring weekly or hourly basis. Ovens feature an electronically controlled damper to vent the work space, a boost feature for rapid heating of cold oven, and a dedicated access port for the introduction of external temperature sensors.
Smooth stainless steel inner chamber has rounded corners for easy cleaning. The door opens 180°, providing easy access to interior for cleaning and loading of probes. An automatic overtemperature alarm ensures your samples are always protected. The RS-232 interface lets you connect to your computer for documentation of parameters.
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Thermo Scientific Heratherm 51028124 AP Mechanical Oven; 2.0 cu ft/Adj Fan/120V 3880048 Mfr # 51028124
COMPARE
2.2 120
$8,199.42CAD / Each
Thermo Scientific Heratherm 51028126 AP Mechanical Oven; 6.4 cu ft/Adj Fan/208-240V 3880052 Mfr # 51028126
COMPARE
6.1 208 / 240
$9,488.63CAD / Each
Thermo Scientific Heratherm 51029329 AP Mechanical Oven; 14 cu ft/Adj Fan/208-240V 3880053 Mfr # 51029329
COMPARE
14 208 / 240
$16,501.97CAD / Each
Thermo Scientific Heratherm 51029343 AP Mechanical Oven; 25.8 cu ft/Adj Fan/208-240V 3880055 Mfr # 51029343
COMPARE
25.8 208 / 240
$20,480.12CAD / Each
Thermo Scientific Heratherm 51029344 APS Mechanical Oven; 25.8 cu ft/SS/208-240V 3880078 Mfr # 51029344
COMPARE
25.8 208 / 240
$22,248.19CAD / Each