Spill-X Bulk Agents Refills

Neutralizes spills for safer cleanup and handling
  • Granular base reduces product waste and disposal costs
  • Speeds spill cleanup to limit worker exposure and minimize production downtime
  • Non-toxic, free-flowing dry chemical is environmentally friendly
  • Easy identification of agent with unique color
These refill agents are available in easy-to-store polypropylene containers. Order to replace specific or multipurpose spill treatment kit containers.
$912.03 - $1,915.41CAD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Chemical Treated Color Description
Availability Pricing
Spill-X 76255 Acid Spill Kit Agent Refill; 50 lb Pail 8629350 Mfr # 76255
COMPARE
Acid Red Acid Spill Kit Agent Refill; 50 lb Pail
$912.03CAD / Each
Spill-X 76396 Caustic Spill Kit Agent Refill; 42 lb Pail 8629352 Mfr # 76396
COMPARE
Caustic Tan Caustic Spill Kit Agent Refill; 42 lb Pail
$1,470.44CAD / Each
Spill-X 78443 Formaldehyde Polymizer Spill Kit Refill; 37 lb Pail 8629356 Mfr # 78443
COMPARE
Formaldehyde Green Formaldehyde Polymizer Spill Kit Refill; 37 lb Pail
$1,915.41CAD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat