Spill-X 76255 Acid Spill Kit Agent Refill; 50 lb Pail

Representative image only.

Spill-X 76255 Acid Spill Kit Agent Refill; 50 lb Pail

Spill-X | Mfr: 76255| Item: 8629350

Neutralizes spills for safer cleanup and handling

$912.03CAD / Each

$912.03CAD / Each

Buy It Together

Current Item

Spill-X 76255 Acid Spill Kit Agent Refill; 50 lb Pail
Spill-X 76255 Acid Spill Kit Agent Refill; 50 lb Pail
8629350

$912.03CAD / Each
+
Spill-X 78776 Acid Spill Treatment/Clean Up Kit
8629200

$1,731.23CAD / Each
=
Total Price 2643.26 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Granular base reduces product waste and disposal costs
  • Speeds spill cleanup to limit worker exposure and minimize production downtime
  • Non-toxic, free-flowing dry chemical is environmentally friendly
  • Easy identification of agent with unique color
  • 1 year warranty

More About this Item

These refill agents are available in easy-to-store polypropylene containers. Order to replace specific or multipurpose spill treatment kit containers.

Economy-size pail of control agent covers a 5-gallon (19 L) spill. Guide and instructions are included with pail for start-to-finish cleanup.

1 Year Warranty
Specifications
  • Chemical TreatedAcid
  • Agent ColorRed
  • ColorRed
  • DescriptionAcid Spill Kit Agent Refill; 50 lb Pail
  • Warranty1 year
Reviews
Q&A