Spill-X 78443 Formaldehyde Polymizer Spill Kit Refill; 37 lb Pail
Representative image only.
Spill-X 78443 Formaldehyde Polymizer Spill Kit Refill; 37 lb Pail
Spill-X | Mfr: 78443| Item: 8629356
Neutralizes spills for safer cleanup and handling
$1,915.41CAD / Each
$1,915.41CAD / Each
Buy It Together
Current Item
Spill-X 78443 Formaldehyde Polymizer Spill Kit Refill; 37 lb Pail
Spill-X 78443 Formaldehyde Polymizer Spill Kit Refill; 37 lb Pail
8629356
$1,915.41CAD / Each
+
Spill-X 78776 Acid Spill Treatment/Clean Up Kit
8629200
$1,731.23CAD / Each
=
Total Price 3646.64 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Granular base reduces product waste and disposal costs
  • Speeds spill cleanup to limit worker exposure and minimize production downtime
  • Non-toxic, free-flowing dry chemical is environmentally friendly
  • Easy identification of agent with unique color
  • 1 year warranty
More About this Item
These refill agents are available in easy-to-store polypropylene containers. Order to replace specific or multipurpose spill treatment kit containers.
Economy-size pail of control agent. Coverage is based on formaldehyde strength. Guide and instructions are included with each pail for start-to-finish cleanup.
1 Year Warranty
Specifications
  • Chemical TreatedFormaldehyde
  • Agent ColorGreen
  • ColorGreen
  • DescriptionFormaldehyde Polymizer Spill Kit Refill; 37 lb Pail