Shimpo Compact Digital Force Gauges
Read results from any angle
  • 1000 Hz sample rate provides faster sampling producing better resolution and more accurate results
  • Ideal for strict tolerances with its high accuracy +/-0.2% F.S.
  • Measure and record compression and tension tests- permits true “push-pull” testing
  • Real time or peak hold mode easily selectable with a single button push
  • 180° reversible display and dual printed keypad allows usage as a portable gauge or mounted upside down on a test stand
  • 1 million operation load cell with superior overload protection provides a long-life, damage free product
  • Rugged, ergonomic die cast aluminum construction allows usage on production floor
  • Variable display response rate aids viewing readings
  • Flexible user preference with one button selection of engineering units
  • Dual power capable with rechargeable battery or AC line voltage
  • CE Mark & RoHS Approval
Digital force gauges feature 180° displays for readable results from any angle. Other features include dual-direction keypad, selectable force direction (push/pull), and selectable power on/off settings. Ergonomically designed gauges fit comfortably in your hand. Rugged all-metal construction gauges are available in a variety of capacities to fit the needs of your application. Universal power operation with included AC adapter/charger (100 to 240 VAC).
$1,024.01 - $1,348.15CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity
Availability Pricing
Shimpo FGE-1XY Compact gauge, 16 oz capacity 9395402 Mfr # FGE-1XY
COMPARE
16 oz
$1,024.01CAD / Each
Shimpo FGE-2XY Compact gauge, 2 lb capacity 9395403 Mfr # FGE-2XY
COMPARE
2 lbs
$1,024.01CAD / Each
Shimpo FGE-10XY Compact gauge, 10 lb capacity 9395406 Mfr # FGE-10XY
COMPARE
10 lbs
$1,024.01CAD / Each
Shimpo FGE-20XY Compact gauge, 20 lb capacity 9395407 Mfr # FGE-20XY
COMPARE
20 lbs
$1,024.01CAD / Each
Shimpo FGE-50XY Compact gauge, 50 lb capacity 9395408 Mfr # FGE-50XY
COMPARE
50 lbs
$1,024.01CAD / Each
Shimpo FGE-100XY Compact gauge, 100 lb capacity 9395409 Mfr # FGE-100XY
COMPARE
100 lbs
$1,024.01CAD / Each
Shimpo FGE-200XY High Capacity Digital Force Gauge, 200 lb 9395413 Mfr # FGE-200XY
COMPARE
200 lb
$1,348.15CAD / Each
Showing 1- 7 of 7