Scienceware Sterileware Sense-able Detectable Scoops, PS
Prevent product recalls
  • Metal and X-ray equipment-detectable material
  • FDA-compliant materials are suitable for food and pharma applications
  • Individual scoop bags are prenotched to open at the handle end to maintain scoop cleanliness
Proprietary FDA-grade polystyrene resin is uniformly distributed throughout the entire scoop to ensure even fragments are detectable. Black color can be easily seen against most contrasting materials. Blue color conforms to industry-recognized standards for metal-detectable materials.
The 2 and 4 oz scoops feature a long-handle design to reduce user contamination and flat bottoms that stay upright for use as weighing boats. Individually gamma sterilized and packaged in polyethylene bags with lot stamps for traceability.
$321.94 - $825.10CAD / Case of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Description
Availability Pricing
Scienceware Sterileware Sense-able Blue Metal Detectable Scoops, Polystyrene, 2 oz; 100/Cs 0720502 Mfr # H36902-0006
COMPARE
60 Sense-able Blue Metal Detectable Scoops, Polystyrene, 2 oz; 100/Cs
$321.94CAD / Case of 100
Scienceware Sterileware Sense-able Blue Metal Detectable Scoops, Polystyrene, 4 oz; 100/Cs 0720504 Mfr # H36904-0006
COMPARE
125 Sense-able Blue Metal Detectable Scoops, Polystyrene, 4 oz; 100/Cs
$478.12CAD / Case of 100
Scienceware Sterileware Sense-able Blue Metal Detectable Scoops, Polystyrene, 8 oz; 100/Cs 0720505 Mfr # H36906-0006
COMPARE
250 Sense-able Blue Metal Detectable Scoops, Polystyrene, 8 oz; 100/Cs
$825.10CAD / Case of 100