Roscid Technologies RO120 Humidity and Temperature Transmitter Accessories
The RO120 series of humidity and temperature transmitters is supported by a range of specialized accessories designed to enhance its functionality and ease of use
  • Field-replaceable humidity board includes a calibration certificate for precise measurements and easy maintenance RO120 display provides real-time data visualization Adapter board facilitates versatile installation options Sample chamber equipped with 1/4" tube fittings Humidity board with barometric pressure sensor includes a calibration certificate, offering combined humidity and barometric pressure readings
Accessories include a field-replaceable humidity board with a calibration certificate, ensuring precise measurements and easy maintenance. The RO120 display provides real-time data visualization specifically for the RO120 humidity probe. An adapter board is also available for the RO120/RO120-B probe. A stainless steel fitting with a 3/4" NPT connection ensures secure connections. Additionally, a sample chamber with 1/4" tube fittings facilitates accurate measurements. For applications requiring barometric pressure readings, a field-replaceable humidity board with a barometric pressure sensor, also including a calibration certificate, is available. These accessories collectively enhance the RO120 series, making it a comprehensive and adaptable solution for various industrial monitoring needs.
$135.99 - $1,268.59CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Roscid Technologies RO120 Display for RO120 Humidity Probe 7893022 Mfr # DIS
COMPARE
RO120 Display for RO120 Humidity Probe
$1,178.71CAD / Each
Roscid Technologies Field Replaceable Humidity Board for RO120 with Calibration Certificate 7893024 Mfr # FRRO
COMPARE
Field Replaceable Humidity Board for RO120 with Calibration Certificate
$934.13CAD / Each
Roscid Technologies Sample Chamber with 1/4" Tube Fittings for RO120 7893025 Mfr # RO120-SC
COMPARE
Sample Chamber with 1/4" Tube Fittings for RO120
$1,268.59CAD / Each
Roscid Technologies Stainless Steel Fitting, 3/4" NPT for RO120 7893026 Mfr # RO120-fitting
COMPARE
Stainless Steel Fitting, 3/4" NPT for RO120
$135.99CAD / Each
Roscid Technologies Field Replaceable Humidity Board for RO120 with Barometric Pressure Sensor and Calibration Certificate 7893027 Mfr # FRROB
COMPARE
Field Replaceable Humidity Board for RO120 with Barometric Pressure Sensor and Calibration Certificate
$1,135.98CAD / Each
Roscid Technologies Adapter Board for RO120/RO120-B Probe Only 7893037 Mfr # RO120-AB
COMPARE
Adapter Board for RO120/RO120-B Probe Only
$135.99CAD / Each