Roscid Technologies Stainless Steel Fitting, 3/4" NPT for RO120
Representative image only.
Roscid Technologies Stainless Steel Fitting, 3/4" NPT for RO120
Roscid Technologies | Mfr: RO120-fitting| Item: 7893026
The RO120 series of humidity and temperature transmitters is supported by a range of specialized accessories designed to enhance its functionality and ease of use
$135.99CAD / Each
$135.99CAD / Each
Product Description
Key Features
  • Field-replaceable humidity board includes a calibration certificate for precise measurements and easy maintenance RO120 display provides real-time data visualization Adapter board facilitates versatile installation options Sample chamber equipped with 1/4" tube fittings Humidity board with barometric pressure sensor includes a calibration certificate, offering combined humidity and barometric pressure readings
More About this Item
Accessories include a field-replaceable humidity board with a calibration certificate, ensuring precise measurements and easy maintenance. The RO120 display provides real-time data visualization specifically for the RO120 humidity probe. An adapter board is also available for the RO120/RO120-B probe. A stainless steel fitting with a 3/4" NPT connection ensures secure connections. Additionally, a sample chamber with 1/4" tube fittings facilitates accurate measurements. For applications requiring barometric pressure readings, a field-replaceable humidity board with a barometric pressure sensor, also including a calibration certificate, is available. These accessories collectively enhance the RO120 series, making it a comprehensive and adaptable solution for various industrial monitoring needs.
Specifications
  • DescriptionStainless Steel Fitting, 3/4" NPT for RO120