Roscid Technologies Field Replaceable Humidity Board for RO120 with Barometric Pressure Sensor and Calibration Certificate
Representative image only.
Roscid Technologies Field Replaceable Humidity Board for RO120 with Barometric Pressure Sensor and Calibration Certificate
Roscid Technologies | Mfr: FRROB| Item: 7893027
The RO120 series of humidity and temperature transmitters is supported by a range of specialized accessories designed to enhance its functionality and ease of use
$1,135.98CAD / Each
$1,135.98CAD / Each
Product Description
Key Features
  • Field-replaceable humidity board includes a calibration certificate for precise measurements and easy maintenance RO120 display provides real-time data visualization Adapter board facilitates versatile installation options Sample chamber equipped with 1/4" tube fittings Humidity board with barometric pressure sensor includes a calibration certificate, offering combined humidity and barometric pressure readings
More About this Item
Accessories include a field-replaceable humidity board with a calibration certificate, ensuring precise measurements and easy maintenance. The RO120 display provides real-time data visualization specifically for the RO120 humidity probe. An adapter board is also available for the RO120/RO120-B probe. A stainless steel fitting with a 3/4" NPT connection ensures secure connections. Additionally, a sample chamber with 1/4" tube fittings facilitates accurate measurements. For applications requiring barometric pressure readings, a field-replaceable humidity board with a barometric pressure sensor, also including a calibration certificate, is available. These accessories collectively enhance the RO120 series, making it a comprehensive and adaptable solution for various industrial monitoring needs.
Specifications
  • DescriptionField Replaceable Humidity Board for RO120 with Barometric Pressure Sensor and Calibration Certificate