Right-To-Know NFPA Notice Chemical Hazard Signs and Kits

Signs and kit to help you with regulatory facility compliance
These signs display the four-color National Fire Protection Association (NFPA) diamond format. Signs feature blank hazard rating panels — write in the information for your specific use.
$15.03 - $53.19CAD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Description
Availability Pricing
Accuform ZFD806VS NFPA Notice Chemical Hazard, Sign Only, 10" X 7", adhesive vinyl 4101006 Mfr # ZFD806VS
COMPARE
Vinyl NFPA Notice Chemical Hazard, Sign Only, 10" X 7", adhesive vinyl
$15.03CAD / Each
Accuform ZFD806VA NFPA Notice Chemical Hazard, Sign Only, 10" X 7", aluminum 4101008 Mfr # ZFD806VA
COMPARE
Aluminum NFPA Notice Chemical Hazard, Sign Only, 10" X 7", aluminum
$27.55CAD / Each
Accuform ZFD837VP NFPA Notice Chemical Hazard, Sign Kit, 14" X 10", plastic 4101010 Mfr # ZFD837VP
COMPARE
Plastic NFPA Notice Chemical Hazard, Sign Kit, 14" X 10", plastic
$53.19CAD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat