Accuform ZFD806VA NFPA Notice Chemical Hazard, Sign Only, 10" X 7", aluminum

Representative image only.

Accuform ZFD806VA NFPA Notice Chemical Hazard, Sign Only, 10" X 7", aluminum

Accuform | Mfr: ZFD806VA| Item: 4101008

Signs and kit to help you with regulatory facility compliance

$27.55CAD / Each

$27.55CAD / Each
Product Description

More About this Item

These signs display the four-color National Fire Protection Association (NFPA) diamond format. Signs feature blank hazard rating panels — write in the information for your specific use.

Specifications
  • HeaderNOTICE
  • LanguagesEnglish
  • MaterialAluminum
  • Width (in)10
  • Height (in)14
  • DescriptionNFPA Notice Chemical Hazard, Sign Only, 10" X 7", aluminum
Reviews
Q&A