Accuform ZFD837VP NFPA Notice Chemical Hazard, Sign Kit, 14" X 10", plastic

Representative image only.

Accuform ZFD837VP NFPA Notice Chemical Hazard, Sign Kit, 14" X 10", plastic

Accuform | Mfr: ZFD837VP| Item: 4101010

Signs and kit to help you with regulatory facility compliance

$53.19CAD / Each

$53.19CAD / Each
Product Description

More About this Item

These signs display the four-color National Fire Protection Association (NFPA) diamond format. Signs feature blank hazard rating panels — write in the information for your specific use.

Kit contains one sign, NFPA wallet guide, and adhesive numbers and hazard symbols which are printed black on clear adhesive polyester to fit the hazard panels.

Specifications
  • HeaderNOTICE
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)10
  • Height (in)14
  • DescriptionNFPA Notice Chemical Hazard, Sign Kit, 14" X 10", plastic
Reviews
Q&A