PYREX VISTA 70024 Class A Graduated Cylinders
Calibrated "to deliver"
Manufactured of Class A borosilicate glass, these metric scale cylinders are enameled with blue graduations for easy reading. Cylinder capacity is in accordance wtih ASTM E-1272. 10mL size has a funnel top for easy filling.
$46.85 - $614.40CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex Vista 70024-10 Graduated Glass Cylinder, 10 mL, to deliver, 1/ea 3450470 Mfr # 70024-10
COMPARE
10 0.1 Glass
$46.85CAD / Each
Pyrex Vista 70024-25 Graduated Glass Cylinder, 25 mL, to deliver, 1/ea 3450471 Mfr # 70024-25
COMPARE
25 0.2 Glass
$62.32CAD / Each
Pyrex Vista 70024-100 Graduated Glass Cylinder, 100 mL, to deliver, 1/ea 3450473 Mfr # 70024-100
COMPARE
100 1 Glass
$75.58CAD / Each
Pyrex Vista 70024-250 Graduated Glass Cylinder, 250 mL, to deliver, 1/ea 3450474 Mfr # 70024-250
COMPARE
250 2 Glass
$99.90CAD / Each
Pyrex Vista 70024-500 Graduated Glass Cylinder, 500 mL, to deliver, 1/ea 3450475 Mfr # 70024-500
COMPARE
500 5 Glass
$137.03CAD / Each
Pyrex Vista 70024-25 Graduated Glass Cylinder, 25 mL, to deliver, 18/cs 3450481 Mfr # 70024-25
COMPARE
25 0.5 Glass
$592.30CAD / Case of 18
Pyrex Vista 70024-50 Graduated Glass Cylinder, 50 mL, to deliver, 18/cs 3450482 Mfr # 70024-50
COMPARE
50 1 Glass
$614.40CAD / Case of 18
Showing 1- 7 of 7