PYREX VISTA 70022 Class A Graduated Cylinders
Calibrated ''to contain''
Manufactured of Class A borosilicate glass, these metric scale cylinders are enameled with blue graduations for easy reading. Cylinder capacity is in accordance wtih ASTM E-1272. 10mL size has a funnel top for easy filling.
$46.85 - $777.95CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex Vista 70022-10 Graduated Glass Cylinder, 10 mL, to contain, 1/ea 3450450 Mfr # 70022-10
COMPARE
10 0.1 Glass
$46.85CAD / Each
Pyrex Vista 70022-25 Graduated Glass Cylinder, 25 mL, to contain, 1/ea 3450451 Mfr # 70022-25
COMPARE
25 0.2 Glass
$64.09CAD / Each
Pyrex Vista 70022-100 Graduated Glass Cylinder, 100 mL, to contain, 1/ea 3450453 Mfr # 70022-100
COMPARE
100 1 Glass
$75.58CAD / Each
Pyrex Vista 70022-250 Graduated Glass Cylinder, 250 mL, to contain, 1/ea 3450454 Mfr # 70022-250
COMPARE
250 2 Glass
$96.21CAD / Each
Pyrex Vista 70022-25 Graduated Glass Cylinder, 25 mL, to contain, 18/cs 3450461 Mfr # 70022-25
COMPARE
25 0.2 Glass
$692.49CAD / Case of 18
Pyrex Vista 70022-50 Graduated Glass Cylinder, 50 mL, to contain, 18/cs 3450462 Mfr # 70022-50
COMPARE
50 1 Glass
$707.23CAD / Case of 18
Pyrex Vista 70022-100 Graduated Glass Cylinder, 100 mL, to contain, 12/cs 3450463 Mfr # 70022-100
COMPARE
100 1 Glass
$489.17CAD / Each
Pyrex Vista 70022-250 Graduated Glass Cylinder, 250 mL, to contain, 12/cs 3450464 Mfr # 70022-250
COMPARE
250 2 Glass
$777.95CAD / Each
Showing 1- 8 of 8