Pyrex 7078 Disposable Glass Serological Pipettes, Sterile, Individually-Wrapped

Serological, glass pipettes come individually wrapped and plugged to insure sterility.
  • Negative graduations for versatiity
  • Fire-polished tips provide burr-free even flow rates
  • Reference: ASTM E-714
These individually-wrapped sterile pipettes are calibrated "to deliver" (TD) with blow out. Long, slender, tapered tips make pipetting faster and easier.
Easy open Steriview® wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size. Colors conform to ISO standards.

NOTE: Pipetting by mouth can be hazardous to your health. We suggest using a mechanical pipetting device.
$913.50 - $1,140.40CAD / Case of 800

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Overall Length (mm)
Availability Pricing
Pyrex 7077-1N Disposable Glass Serological Pipette, 1/0.01 mL, Sterile-Indv; 800/Cs 3456622 Mfr # 7077-1N
COMPARE
1 290
$913.50CAD / Case of 800
Pyrex 7077-2N Disposable Glass Serological Pipette, 2 mL, Sterile-Indv; 720/Cs 3456623 Mfr # 7077-2N
COMPARE
2 290
$969.49CAD / Case of 720
Pyrex 7077-5N Disposable Glass Serological Pipette, 5 mL, Sterile-Indv; 720/Cs 3456624 Mfr # 7077-5N
COMPARE
5 290
$1,140.40CAD / Case of 720
Pyrex 7077-10N Disposable Glass Serological Pipette, 10 mL, Sterile-Indv; 600/Cs 3456625 Mfr # 7077-10N
COMPARE
10 290
$1,065.26CAD / Case of 600
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat