Pyrex 7077-2N Disposable Glass Serological Pipette, 2 mL, Sterile-Indv; 720/Cs

Representative image only.

Pyrex 7077-2N Disposable Glass Serological Pipette, 2 mL, Sterile-Indv; 720/Cs

Pyrex | Mfr: 7077-2N| Item: 3456623

Serological, glass pipettes come individually wrapped and plugged to insure sterility.

$969.49CAD / Case of 720

$969.49CAD / Case of 720
Product Description

Key Features

  • Negative graduations for versatiity
  • Fire-polished tips provide burr-free even flow rates
  • Reference: ASTM E-714

More About this Item

These individually-wrapped sterile pipettes are calibrated "to deliver" (TD) with blow out. Long, slender, tapered tips make pipetting faster and easier.
Easy open Steriview® wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size. Colors conform to ISO standards.

NOTE: Pipetting by mouth can be hazardous to your health. We suggest using a mechanical pipetting device.

Specifications
  • SterileYes
  • AutoclavableYes
  • Max Volume (µL)2000
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)2
  • Max Dispensing (mL)2
  • Graduations (mL)0.01
  • Overall Length (InchShort)11.41
  • Overall Length (mm)290
  • Tip StyleStandard
  • Body MaterialGlass
  • Color CodeGreen
  • CalibrationTo Deliver (TD)
  • Package OfIndividually wrapped (Steri-View wrapper)
  • DescriptionDisposableGlassSerologicalPipette,2mL,Sterile-Indv;720/Cs
Reviews
Q&A