Pyrex 7077-1N Disposable Glass Serological Pipette, 1/0.01 mL, Sterile-Indv; 800/Cs

Representative image only.

Pyrex 7077-1N Disposable Glass Serological Pipette, 1/0.01 mL, Sterile-Indv; 800/Cs

Pyrex | Mfr: 7077-1N| Item: 3456622

Serological, glass pipettes come individually wrapped and plugged to insure sterility.

$913.50CAD / Case of 800

$913.50CAD / Case of 800
Product Description

Key Features

  • Negative graduations for versatiity
  • Fire-polished tips provide burr-free even flow rates
  • Reference: ASTM E-714

More About this Item

These individually-wrapped sterile pipettes are calibrated "to deliver" (TD) with blow out. Long, slender, tapered tips make pipetting faster and easier.
Easy open Steriview® wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size. Colors conform to ISO standards.

NOTE: Pipetting by mouth can be hazardous to your health. We suggest using a mechanical pipetting device.

Specifications
  • SterileYes
  • AutoclavableYes
  • Max Volume (µL)1000
  • Min Volume (µL)10
  • Min Volume (mL)0.01
  • Max Volume (mL)1
  • Max Dispensing (mL)1
  • Graduations (mL)0.01
  • Overall Length (InchShort)11.41
  • Overall Length (mm)290
  • Tip StyleStandard
  • Body MaterialGlass
  • Color CodeYellow
  • CalibrationTo Deliver (TD)
  • Package OfIndividually wrapped (Steri-View wrapper)
  • DescriptionDisposableGlassSerologicalPipette,1/0.01mL,Sterile-Indv;800/Cs
Reviews
Q&A