METTLER TOLEDO SevenGo Duo Series Handheld Multiparameter Meters
Rugged, portable, and ergonomic
  • Ergonomic design provides easy right-or left-handed operation
  • IP67-rated housing is watertight and dustproof, including sensor connections
  • Automatic end point, sensor, and buffer recognition functions simplify operation
$1,900.67 - $5,024.26CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo SG23 Handheld pH / Conductivity Meter Only 3590370 Mfr # 51302600
COMPARE
Handheld pH / Conductivity Meter Only
$1,900.67CAD / Each
Mettler Toledo SG23 Handheld pH / Conductivity Meter Standard Kit 3590371 Mfr # 51302601
COMPARE
Handheld pH / Conductivity Meter Standard Kit
$3,013.08CAD / Each
Mettler Toledo SG23 Handheld pH / Conductivity Meter Field Kit 3590372 Mfr # 51302602
COMPARE
Handheld pH / Conductivity Meter Field Kit
$3,322.49CAD / Each
Mettler Toledo SG68 Handheld pH / Ion / DO Meter Only 3590373 Mfr # 51302610
COMPARE
Handheld pH / Ion / DO Meter Only
$3,514.04CAD / Each
Mettler Toledo SG68 Handheld pH / Ion / DO Meter Standard Kit 3590374 Mfr # 51302611
COMPARE
Handheld pH / Ion / DO Meter Standard Kit
$5,024.26CAD / Each
Mettler Toledo SG78 Handheld pH / Ion / Conductivity Meter Standard Kit 3590377 Mfr # 51302621
COMPARE
Handheld pH / Ion / Conductivity Meter Standard Kit
$4,096.02CAD / Each