Mettler Toledo SG68 Handheld pH / Ion / DO Meter Standard Kit

Representative image only.

Mettler Toledo SG68 Handheld pH / Ion / DO Meter Standard Kit

Mettler Toledo | Mfr: 51302611| Item: 3590374

Rugged, portable, and ergonomic

$5,024.26CAD / Each

$5,024.26CAD / Each
Product Description

Includes

InLab® Expert Pro-ISM pH electrode and InLab 605-ISM polarographic dissolved oxygen probe, both with IP67 and 6-ft (1.8-m) cables, wrist strap, 4 AA batteries, and test certificate supplied by the manufacturer.

Key Features

  • Ergonomic design provides easy right-or left-handed operation
  • IP67-rated housing is watertight and dustproof, including sensor connections
  • Automatic end point, sensor, and buffer recognition functions simplify operation
  • 1 year warranty

More About this Item

The SevenGo Duo Pro handheld multiparameter meter displays both measured parameters simultaneously. All measurements are stored with time-and-date stamp, electrode and sample data, and other relevant information to ensure GLP guidelines are met. Download saved data to PC via RS-232 or USB (order adapter cables separately).

1 Year Warranty
Specifications
  • Min pH (pH)-2
  • Max pH (pH)19.999
  • pH Resolution0.001
  • pH Accuracy±0.002
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1
  • mV Accuracy±0.1
  • Min ISE0
  • Max ISE9999 ppm
  • ISE Resolution3 or 4 digits
  • ISE Accuracy±0.5%
  • Min Dissolved Oxygen0
  • Max Dissolved Oxygen99 ppm
  • Dissolved Oxygen Resolution0.01
  • Dissolved Oxygen Accuracy±0.5% max. 0.03
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.2°C
  • Temperature compensation-5 to 130°C (ATC), -30 to 130°C (MTC)
  • OutputUSB, RS-232, Infrared
  • Data Logging (points)500 measurements (GLP)
  • WaterproofYes
  • Battery4 AA (included)
  • Length (cm)22
  • Width (cm)9
  • Height (cm)4.5
  • Width (in)3 1/2
  • Length (in)8 11/16
  • Height (in)1 3/4
  • DescriptionHandheld pH / Ion / DO Meter Standard Kit
  • Warranty1 year
Reviews
Q&A