Mettler Toledo SG23 Handheld pH / Conductivity Meter Field Kit

Representative image only.

Mettler Toledo SG23 Handheld pH / Conductivity Meter Field Kit

Mettler Toledo | Mfr: 51302602| Item: 3590372

Rugged, portable, and ergonomic

$3,322.49CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$3,322.49CAD / Each

Buy It Together

Current Item

Mettler Toledo SG23 Handheld pH / Conductivity Meter Field Kit
Mettler Toledo SG23 Handheld pH / Conductivity Meter Field Kit
3590372

$3,322.49CAD / Each
+ =
Total Price 3596.17 ADD 2 ITEMS TO CART
Product Description

Includes

InLab® Expert Pro-ISM pH electrode and InLab 738-ISM conductivity cell, both with IP67 and 6-ft (1.8-m) cables, uGo™ carrying case, wrist strap, 4 AA batteries, and test certificate supplied by the manufacturer.

Key Features

  • Ergonomic design provides easy right-or left-handed operation
  • IP67-rated housing is watertight and dustproof, including sensor connections
  • Automatic end point, sensor, and buffer recognition functions simplify operation
  • 1 year warranty

More About this Item

The SevenGo Duo handheld multiparameter meter measures each channel simultaneously and the large screen toggles between the parallel readings.

1 Year Warranty
Specifications
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01
  • pH Accuracy±0.01
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1
  • mV Accuracy±1
  • Min Conductivity0.01 µS/cm
  • Max Conductivity500 mS/cm
  • Conductivity Resolution0.01 to 1
  • Conductivity Accuracy±0.01%
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.5°C
  • Temperature compensation-5 to 105°C
  • Data Logging (points)99 measurements
  • WaterproofYes
  • Battery4 AA (included)
  • Length (cm)22
  • Width (cm)9
  • Height (cm)4.5
  • Width (in)3 1/2
  • Length (in)8 11/16
  • Height (in)1 3/4
  • DescriptionHandheld pH / Conductivity Meter Field Kit
  • Warranty1 year
Reviews
Q&A