Mettler Toledo S8 Handheld pH/Ion Meter Field Kit

Representative image only.

Mettler Toledo S8 Handheld pH/Ion Meter Field Kit

Mettler Toledo | Mfr: 30207877| Item: 3590363

No more juggling while multitasking

$2,055.38CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$2,055.38CAD / Each

Buy It Together

Current Item

Mettler Toledo S8 Handheld pH/Ion Meter Field Kit
Mettler Toledo S8 Handheld pH/Ion Meter Field Kit
3590363

$2,055.38CAD / Each
+ + =
Total Price 3631.39 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Inlab Expert Pro-ISM-IP67 pH electrode, buffer sachet set, solution bottles, carrying case, LabX direct pH software, USB cable, electrode holder, stabilizing/mounting base, wrist strap, test report, and 4 AA batteries.

Key Features

  • Single-handed operation frees a hand for other tasks
  • Robust IP67-rated housing protects meter from harsh conditions
  • Enhanced contrast screen is readable in any light condition—even bright sunlight
  • Compatible with InLab® ISM sensors for quick electrode changes without recalibration
  • 1 year warranty

More About this Item

This handheld meter withstands the rigors of busy analytical laboratories, demanding process environments, and unpredictable outdoor applications. The directional keypad with central enter key and intuitive simplified interface allow one hand to set up, calibrate, and measure while the other hand remains free. Waterproof housing resists washdown exposure and inclement weather.

Choose the kit with an electrode and accessories that best fit your application.

The InLab Expert Pro-ISM pH electrode is ideal for general applications in or out of doors, with a rugged PEEK body and double junction to protect the sensor from dirty samples.

1 Year Warranty
Specifications
  • Meter TypePortable
  • Display typeGraphic LCD
  • ConnectionsMicro-USB
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001, 0.01, 0.1
  • pH Accuracy±0.002
  • Buffer RecognitionNIST, DIN, JIS, MERCK, MT USA, MT Europe, JJG 119, Technical, Custom
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1, 1
  • mV Accuracy±0.1
  • Min ISEDepends on the electrode used
  • Max ISEDepends on the electrode used
  • ISE Resolution3 or 4 digits (auto range)
  • ISE Accuracy±0.5%
  • Min Temperature (° C)-5
  • Max Temperature (° C)130
  • Min Temperature (° F)23
  • Max Temperature (° F)266
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.2° up to 105°C, ±0.5° above 105°C
  • Temperature compensationAutomatic or manual
  • Data Logging (points)2000 with time and date
  • WaterproofYes
  • Battery4 AA (included)
  • Battery life (hr)250
  • Length (cm)22.2
  • Width (cm)7
  • Height (cm)3.5
  • Width (in)2 3/4
  • Length (in)8 3/4
  • Height (in)1 3/8
  • DescriptionHandheld pH/Ion Meter Field Kit
  • Warranty1 year
Reviews
Q&A